Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srr
Entrez Gene ID
27364
Gene Name
serine racemase
Gene Aliases
Srs
Chromosome Location
Chr.11: 74905047 - 74925798 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163311.1 NP_001156783.1 5 - 6 702 80
NM_013761.4 NP_038789.1 6 - 7 928 80
GenBank mRNA AF148321.1 - 5 - 6 583 80
AK017235.1 - 6 - 7 807 80
AK028034.1 - 5 - 6 585 80
AK043738.1 - 6 - 7 834 80
AK079343.1 - 2 - 3 212 80
AK157122.1 - 5 - 6 853 80
AK170096.1 - 6 - 7 814 80
AK170471.1 - 6 - 7 814 80
BC011164.1 - 6 - 7 805 80

View Full Product Details