Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erbb3
Entrez Gene ID
13867
Gene Name
v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Aliases
C76256, Erbb-3, Erbb3r, Her3
Chromosome Location
Chr.10: 128569368 - 128589501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010153.1 NP_034283.1 20 - 21 2448 67
GenBank mRNA AY686636.1 - 20 - 21 2448 67
BC036180.1 - - 52 67
BC049279.1 - - 260 67
BC106091.1 - - 2599 67

View Full Product Details