Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stx1a
Entrez Gene ID
20907
Gene Name
syntaxin 1A (brain)
Gene Aliases
HPC-1
Chromosome Location
Chr.5: 135023487 - 135051104 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016801.3 NP_058081.2 2 - 3 116 60
GenBank mRNA BC047133.1 - 2 - 3 105 60
BC057892.1 - 2 - 3 128 60
BC100446.1 - 2 - 3 151 60
D45208.2 - 2 - 3 116 60

View Full Product Details