The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040691.1 | NP_001035781.1 | 1 - 2 | 192 | 59 |
GenBank mRNA | AK146178.1 | - | 1 - 2 | 192 | 59 |
BC011039.1 | - | 1 - 2 | 249 | 59 | |
Y08975.1 | - | 1 - 2 | 192 | 59 |