Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspa4
Entrez Gene ID
15525
Gene Name
heat shock protein 4
Gene Aliases
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025
Chromosome Location
Chr.11: 53259814 - 53300479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008300.3 NP_032326.3 16 - 17 2357 92
GenBank mRNA AK133772.1 - 16 - 17 2300 92
AK148883.1 - 5 - 6 632 92
AK155305.1 - 16 - 17 2274 92
AK166102.1 - 16 - 17 2340 92
AK220167.1 - 16 - 17 2308 92
AW545353.1 - 3 - 4 92
BC003770.1 - 16 - 17 2263 92
BC003950.1 - 16 - 17 2250 92
CA316420.1 - 5 - 6 652 92
D85904.1 - 16 - 17 2056 92

View Full Product Details