The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008300.3 | NP_032326.3 | 18 - 19 | 2638 | 98 |
GenBank mRNA | AK133772.1 | - | 18 - 19 | 2581 | 98 |
AK155305.1 | - | 18 - 19 | 2555 | 98 | |
AK166102.1 | - | 18 - 19 | 2621 | 98 | |
AK220167.1 | - | 18 - 19 | 2589 | 98 | |
AW545353.1 | - | 1 - 2 | 98 | ||
BC003770.1 | - | 18 - 19 | 2544 | 98 | |
BC003950.1 | - | 18 - 19 | 2531 | 98 | |
D85904.1 | - | 18 - 19 | 2337 | 98 |