The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011075.2 | NP_035205.1 | 3 - 4 | 260 | 108 |
GenBank mRNA | AK030328.1 | - | 3 - 4 | 261 | 108 |
BC141363.1 | - | 3 - 4 | 217 | 108 | |
BC150811.1 | - | 3 - 4 | 217 | 108 | |
M14757.1 | - | 3 - 4 | 220 | 108 |