Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRM2B
Entrez Gene ID
101123639
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Gene Aliases
-
Chromosome Location
Chr.9: 82961003 - 82997280 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012184037.2 XP_012039427.2 2 - 3 325 68
XM_012184039.3 XP_012039429.2 2 - 3 330 68
XM_012184040.2 XP_012039430.2 2 - 3 332 68
XM_027973160.1 XP_027828961.1 2 - 3 325 68
XM_027973161.1 XP_027828962.1 2 - 3 327 68

View Full Product Details