Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBX6
Entrez Gene ID
101115502
Gene Name
T-box transcription factor 6
Gene Aliases
-
Chromosome Location
Chr.24: 26732333 - 26737101 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962047.1 XP_027817848.1 4 - 5 1482 81

View Full Product Details