Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGT1A7
Entrez Gene ID
100037707
Gene Name
UDP glucuronosyltransferase 1 family, polypeptide A7
Gene Aliases
UGT1A8
Chromosome Location
Chr.Unknown: 1153163 - 1282924 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195629.1 NP_001182558.1 - 850 113
GenBank mRNA AF011902.1 - - 850 113

View Full Product Details