Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX6
Entrez Gene ID
710353
Gene Name
T-box transcription factor 6
Gene Aliases
-
Chromosome Location
Chr.20: 27264113 - 27270560 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015125907.2 XP_014981393.1 - 1359 76
XM_028840862.1 XP_028696695.1 - 978 76

View Full Product Details