Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED10
Entrez Gene ID
694872
Gene Name
mediator complex subunit 10
Gene Aliases
EGK_16344
Chromosome Location
Chr.6: 6287158 - 6293743 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260853.1 NP_001247782.1 - 259 96
GenBank mRNA JU336686.1 - - 254 96
JU477482.1 - - 259 96

View Full Product Details