Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXN2
Entrez Gene ID
715878
Gene Name
forkhead box N2
Gene Aliases
EGK_05314
Chromosome Location
Chr.13: 60361935 - 60427269 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261043.1 NP_001247972.1 - 645 139
XM_015112307.2 XP_014967793.1 - 834 139
XM_015112309.2 XP_014967795.1 - 968 139
XM_015112310.2 XP_014967796.1 - 734 139
XM_015112311.2 XP_014967797.1 - 732 139
XM_028831621.1 XP_028687454.1 - 1011 139
XM_028831622.1 XP_028687455.1 - 1193 139
XM_028831623.1 XP_028687456.1 - 1206 139
XM_028831624.1 XP_028687457.1 - 717 139
GenBank mRNA JU334307.1 - - 734 139
JU334308.1 - - 734 139
JU475413.1 - - 645 139
JU475414.1 - - 645 139

View Full Product Details