The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012634.1 | NP_036766.1 | 1 - 2 | 469 | 64 |
XM_006256829.2 | XP_006256891.1 | 1 - 2 | 232 | 64 | |
GenBank mRNA | M17259.1 | - | 1 - 2 | 210 | 64 |
M29393.1 | - | 1 - 2 | 210 | 64 | |
X16555.1 | - | 1 - 2 | 469 | 64 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006256848.2 | XP_006256910.2 | 1 - 2 | 319 | 64 |
XM_008773204.1 | XP_008771426.1 | - | 236 | 64 |