The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053430.1 | NP_445882.1 | 1 - 2 | 243 | 62 |
GenBank mRNA | AF281018.1 | - | 1 - 2 | 243 | 62 |