The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053788.2 | NP_446240.2 | 1 - 2 | 61 | 70 |
GenBank mRNA | AF217191.1 | - | 1 - 2 | 30 | 70 |
D10392.1 | - | 1 - 2 | 61 | 70 |