Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec3a
Entrez Gene ID
365009
Gene Name
C-type lectin domain family 3, member A
Gene Aliases
Clecsf1
Chromosome Location
Chr.19: 46733668 - 46741245 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108899.1 NP_001102369.1 1 - 2 119 82

View Full Product Details