Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Polr3g
Entrez Gene ID
685465
Gene Name
polymerase (RNA) III (DNA directed) polypeptide G
Gene Aliases
-
Chromosome Location
Chr.2: 9537642 - 9578551 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109468.1 NP_001102938.1 3 - 4 940 97
XM_006231729.2 XP_006231791.1 2 - 3 282 97
GenBank mRNA FQ209718.1 - 3 - 4 699 97
FQ210837.1 - 3 - 4 699 97

View Full Product Details