Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajc9
Entrez Gene ID
364240
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 9
Gene Aliases
JDD1
Chromosome Location
Chr.15: 4356512 - 4360788 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108865.1 NP_001102335.1 4 - 5 760 134
GenBank mRNA AB002591.1 - 4 - 5 1011 134
FQ215794.1 - 4 - 5 134
FQ229094.1 - 4 - 5 742 134

View Full Product Details