Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc22
Entrez Gene ID
298300
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
RGD1561518
Chromosome Location
Chr.5: 126225529 - 126243178 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106671.1 NP_001100141.1 6 - 7 1273 61
XM_006238502.2 XP_006238564.1 3 - 4 1825 61

View Full Product Details