The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011993.1 | NP_001011993.1 | 4 - 5 | 612 | 76 |
GenBank mRNA | BC087166.1 | - | 4 - 5 | 612 | 76 |