Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DLT1
Entrez Gene ID
855156
Gene Name
Gene Aliases
YMR126C
Chromosome Location
Chr.XIII: 520761 - 521789 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182627.1 NP_013845.1 1 - 1 951 85

View Full Product Details