Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLZ1
Entrez Gene ID
855525
Gene Name
Gene Aliases
YNL196C
Chromosome Location
Chr.XIV: 270275 - 271171 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183034.1 NP_014203.1 1 - 1 744 82

View Full Product Details