Gene Transcripts
- Gene Symbol
- NFATC3
- Entrez Gene ID
- 4775
- Gene Name
- nuclear factor of activated T-cells 3
- Gene Aliases
- NFAT4, NFATX
- Chromosome Location
- Chr. 16: 68085366 - 68229259 on Build GRCh38
- UniGene ID
- Hs.436585
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM700821.1 |
- |
1 |
53 |
|
Chr. 16 - 68227942 - 68228502 [+] |
GenBankmRNA |
BQ425025.1 |
- |
1 |
408 |
|
Chr. 16 - 68227586 - 68228434 [+] |
GenBankmRNA |
CR607695.1 |
- |
1 |
761 |
|
Chr. 16 - 68227226 - 68229145 [+] |
GenBankmRNA |
CX873126.1 |
- |
1 |
272 |
|
Chr. 16 - 68227715 - 68228430 [+] |
View Full Product Details