Gene Transcripts

Gene Symbol
Olfml1
Entrez Gene ID
244198
Gene Name
olfactomedin-like 1
Gene Aliases
6720478C22, AV321997, BC047207, MVAL564, ONT2, mONT2
Chromosome Location
Chr. 7: 107567433 - 107591365 on Build GRCm38
UniGene ID
Mm.77425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172907.3 NP_766495.1 2 496
GenBank mRNA AK032948.1 - 2 457
AK140277.1 - 2 249
BC047207.1 - 2 483 6310165

View Full Product Details