Gene Transcripts

Gene Symbol
Trim65
Entrez Gene ID
338364
Gene Name
tripartite motif-containing 65
Gene Aliases
4732463G12Rik
Chromosome Location
Chr. 11: 116124708 - 116131128 on Build GRCm38
UniGene ID
Mm.439976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178802.4 NP_848917.2 5, 6 1127
GenBank mRNA AK028857.1 - 5, 6 1016
AK047439.1 - 5, 6 1127
AK050080.1 - 5, 6 1039
AK079982.1 - 4, 5 1209
BC057094.1 - 1016 6418691
BC118954.1 - 5, 6 882
BC118955.1 - 5, 6 882

View Full Product Details