Gene Transcripts

Gene Symbol
Myl9
Entrez Gene ID
296313
Gene Name
myosin, light chain 9, regulatory
Gene Aliases
Chromosome Location
Chr. 3: 152857575 - 152863961 on Build Rnor_6.0
UniGene ID
Rn.6870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100885.1 NP_001094355.1 1 142
XM_006235401.1 XP_006235463.1 2 258
XM_006235402.2 XP_006235464.1 2 272
GenBank mRNA BC158548.1 - 2 221
FQ215756.1 - 2 195
FQ219977.1 - 2 199
FQ220245.1 - 2 199
FQ220303.1 - 2 199
FQ220687.1 - 2 201
FQ227515.1 - 2 202
FQ229420.1 - 2 199
FQ229727.1 - 2 202
FQ229762.1 - 2 201
FQ230056.1 - 2 202
FQ230407.1 - 2 215
FQ231493.1 - 2 215
FQ231820.1 - 2 234
FQ232091.1 - 2 195
S77900.1 - 1 142

View Full Product Details