Gene Transcripts

Gene Symbol
Msc
Entrez Gene ID
312897
Gene Name
musculin
Gene Aliases
Chromosome Location
Chr. 5: 3954902 - 3956974 on Build Rnor_6.0
UniGene ID
Rn.110583
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191754.1 NP_001178683.1 1 497
XM_008763476.1 XP_008761698.1 1 822

View Full Product Details