Gene Transcripts

Gene Symbol
Spetex-2B
Entrez Gene ID
497084
Gene Name
Spetex-2B protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011697.1 NP_001011697.1 2 158
GenBank mRNA AB180077.1 - 2 158

Gene Symbol
Spetex-2A
Entrez Gene ID
364261
Gene Name
Spetex-2A protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009976.1 NP_001009976.1 2 203
GenBank mRNA AB180076.1 - 2 203

Gene Symbol
Spetex-2C
Entrez Gene ID
364255
Gene Name
Spetex-2C protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.215809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011698.1 NP_001011698.1 2 158
GenBank mRNA AB180078.1 - 2 158

Gene Symbol
Spetex-2D
Entrez Gene ID
364260
Gene Name
Spetex-2D protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011701.1 NP_001011701.1 2 158
GenBank mRNA AB180079.1 - 2 158

View Full Product Details