Gene Transcripts

Gene Symbol
Cers3
Entrez Gene ID
499174
Gene Name
ceramide synthase 3
Gene Aliases
Lass3, RGD1564356
Chromosome Location
Chr. 1: 127706618 - 127781041 on Build Rnor_6.0
UniGene ID
Rn.130397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127561.1 NP_001121033.1 306
GenBank mRNA BC160913.1 - 306

View Full Product Details