Gene Transcripts

Gene Symbol
UBE2U
Entrez Gene ID
148581
Gene Name
ubiquitin conjugating enzyme E2 U (putative)
Gene Aliases
Chromosome Location
Chr. 1: 64202802 - 64267381 on Build GRCh38
UniGene ID
Hs.388746
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152489.1 NP_689702.1 4, 5 591
XM_005270498.4 XP_005270555.1 4, 5 1385
XM_006710379.3 XP_006710442.1 4, 5 526
XM_011540764.2 XP_011539066.1 5, 6 1326
GenBank mRNA BC029895.1 - 4, 5 591

View Full Product Details