Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023202.4 | NP_075691.1 | 3 | 253 | |
GenBank mRNA | AK002476.1 | - | 3 | 234 | |
AK003468.1 | - | 3 | 234 | ||
AK010299.1 | - | 3 | 211 | ||
BC052817.1 | - | 3 | 217 | 6487401 | |
BC055698.1 | - | 3 | 218 | 6411262 | |
BX518947.1 | - | 2 | 130 | ||
KJ650204.1 | - | 3 | 195 |