Gene Transcripts
- Gene Symbol
- Vmn1r25
- Entrez Gene ID
- 113865
- Gene Name
- vomeronasal 1 receptor 25
- Gene Aliases
- V1rc8
- Chromosome Location
- Chr. 6: 57978394 - 57979302 on Build GRCm38
- UniGene ID
- Mm.431972
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053238.2 |
NP_444468.2
|
1 |
692 |
|
View Full Product Details