Gene Transcripts

Gene Symbol
Angptl6
Entrez Gene ID
70726
Gene Name
angiopoietin-like 6
Gene Aliases
6330404E11Rik, AGF, ARP3, Arp5
Chromosome Location
Chr. 9: 20873808 - 20879816 on Build GRCm38
UniGene ID
Mm.34019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145154.2 NP_660136.1 3 769
GenBank mRNA AB054065.1 - 3 769
AK018113.1 - 2 138
BC025904.1 - 3 747 5049765

View Full Product Details