Gene Transcripts

Gene Symbol
SCYL1
Entrez Gene ID
57410
Gene Name
SCY1 like pseudokinase 1
Gene Aliases
GKLP, HT019, NKTL, NTKL, P105, SCAR21, TAPK, TEIF, TRAP
Chromosome Location
Chr. 11: 65525065 - 65538711 on Build GRCh38
UniGene ID
Hs.238839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048218.1 NP_001041683.1 8 1117
NM_020680.3 NP_065731.3 8 1117
XM_005274118.3 XP_005274175.1 8 1129
XM_005274120.3 XP_005274177.1 8 1129
XM_005274121.3 XP_005274178.1 8 1129
XM_017018029.1 XP_016873518.1 8 1129
XM_017018030.1 XP_016873519.1 8 1129
XM_017018031.1 XP_016873520.1 8 1129
XM_017018032.1 XP_016873521.1 8 971
XM_017018033.1 XP_016873522.1 8 1121
GenBank mRNA AB047077.1 - 8 1040
AB051427.1 - 8 1040
AB051428.1 - 8 1040
AF225424.1 - 2 181
AF297709.1 - 8 1079
AK314945.1 - 8 1073
BC009967.2 - 2 115
BC069233.1 - 8 1081 6581110

View Full Product Details