Gene Transcripts

Gene Symbol
IL4I1
Entrez Gene ID
259307
Gene Name
interleukin 4 induced 1
Gene Aliases
FIG1, LAAO, LAO
Chromosome Location
Chr. 19: 49889654 - 49929539 on Build GRCh38
UniGene ID
Hs.574492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258017.1 NP_001244946.1 10 2301
NM_001258018.1 NP_001244947.1 10 2383
NM_152899.1 NP_690863.1 8 1774
NM_172374.2 NP_758962.1 10 2325
NR_047577.1 11 2360
GenBank mRNA AF293462.1 - 8 1774
AJ880386.1 - 10 2200
AJ880387.1 - 10 2177
AJ880388.1 - 10 2239
AJ880389.1 - 11 2237
AY358933.1 - 8 1774
BC026103.1 - 10 2229
BC064378.1 - 8 1760 5725335
BC090852.1 - 8 1761 6191892
BC131625.1 - 10 2214 40117891
DQ079587.1 - 10 2325
DQ079588.1 - 10 2383
DQ079589.1 - 10 2301

View Full Product Details