Gene Transcripts
- Gene Symbol
- ABCF3
- Entrez Gene ID
- 55324
- Gene Name
- ATP binding cassette subfamily F member 3
- Gene Aliases
- EST201864
- Chromosome Location
- Chr. 3: 184186075 - 184194012 on Build GRCh38
- UniGene ID
- Hs.361323
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018358.2 |
NP_060828.2
|
3, 4 |
431 |
|
GenBank mRNA |
AK002060.1 |
- |
3, 4 |
408 |
|
AK027288.1 |
- |
3, 4 |
306 |
|
AK092415.1 |
- |
3, 4 |
314 |
|
AK097073.1 |
- |
2, 3 |
471 |
|
AK290106.1 |
- |
3, 4 |
303 |
|
AK299256.1 |
- |
3, 4 |
310 |
|
AK299433.1 |
- |
3, 4 |
195 |
|
BC009253.2 |
- |
3, 4 |
310 |
3028775 |
BC035652.1 |
- |
3, 4 |
305 |
|
BC051754.2 |
- |
3, 4 |
305 |
6064628 |
BC051884.1 |
- |
3, 4 |
305 |
5105761 |
DB079392.1 |
- |
3, 4 |
363 |
|
DC318082.1 |
- |
3, 4 |
483 |
|
KU178694.1 |
- |
3, 4 |
298 |
|
KU178695.1 |
- |
3, 4 |
298 |
|
View Full Product Details