Gene Transcripts

Gene Symbol
SPATA5
Entrez Gene ID
166378
Gene Name
spermatogenesis associated 5
Gene Aliases
AFG2, EHLMRS, SPAF
Chromosome Location
Chr. 4: 122923070 - 123319450 on Build GRCh38
UniGene ID
Hs.709648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317799.1 NP_001304728.1 5 1024
NM_145207.2 NP_660208.2 5 1027
XM_005262783.4 XP_005262840.1 5 1024
XM_011531678.2 XP_011529980.1 5 1024
XM_011531679.2 XP_011529981.1 5 982
XM_017007825.1 XP_016863314.1 5 1023
XM_017007826.1 XP_016863315.1 5 1023
XM_017007827.1 XP_016863316.1 5 1023
XM_017007828.1 XP_016863317.1 5 989
XM_017007829.1 XP_016863318.1 3 1647
XM_017007830.1 XP_016863319.1 5 1023
GenBank mRNA AF361489.1 - 5 1023
AF479656.1 - 5 997
AK091384.1 - 5 982
AL832175.1 - 5 1020
BC048217.1 - 5 967 6052305

View Full Product Details