Gene Transcripts
- Gene Symbol
- CDC25C
- Entrez Gene ID
- 995
- Gene Name
- cell division cycle 25C
- Gene Aliases
- CDC25, PPP1R60
- Chromosome Location
- Chr. 5: 138285265 - 138338355 on Build GRCh38
- UniGene ID
- Hs.656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287582.1 |
NP_001274511.1
|
7, 8 |
935 |
|
NM_001287583.1 |
NP_001274512.1
|
7, 8 |
1052 |
|
NM_001318098.1 |
NP_001305027.1
|
7, 8 |
1056 |
|
NM_001790.4 |
NP_001781.2
|
7, 8 |
939 |
|
NM_022809.3 |
NP_073720.1
|
4, 5 |
720 |
|
XM_005272145.3 |
XP_005272202.1
|
7, 8 |
1052 |
|
XM_006714739.3 |
XP_006714802.1
|
6, 7 |
650 |
|
XM_011543759.2 |
XP_011542061.1
|
8, 9 |
723 |
|
XM_011543760.2 |
XP_011542062.1
|
7, 8 |
1052 |
|
XM_011543761.2 |
XP_011542063.1
|
4, 5 |
833 |
|
XM_011543762.1 |
XP_011542064.1
|
4, 5 |
633 |
|
XM_011543763.1 |
XP_011542065.1
|
5, 6 |
348 |
|
XM_011543764.1 |
XP_011542066.1
|
4, 5 |
401 |
|
XM_017010101.1 |
XP_016865590.1
|
7, 8 |
930 |
|
GenBank mRNA |
AF277723.1 |
- |
|
435 |
|
AF277724.1 |
- |
|
401 |
|
AF277725.1 |
- |
|
396 |
|
AF277726.1 |
- |
|
306 |
|
AF312681.1 |
- |
2, 3 |
182 |
|
AJ304504.1 |
- |
3, 4 |
399 |
|
AK097710.1 |
- |
7, 8 |
1052 |
|
AK301828.1 |
- |
8, 9 |
1002 |
|
BC019089.2 |
- |
7, 8 |
829 |
4858949 |
M34065.1 |
- |
7, 8 |
828 |
|
View Full Product Details