Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042463.2 | NP_001035928.2 | 5 | 1445 | |
NM_001276253.1 | NP_001263182.1 | 4 | 599 | ||
NM_174940.3 | NP_777600.3 | 5 | 1421 | ||
XM_006718206.3 | XP_006718269.3 | 6 | 798 | ||
XM_017017600.1 | XP_016873089.1 | 6 | 973 | ||
GenBank mRNA | AK124309.1 | - | 5 | 1852 | |
BC008671.1 | - | 5 | 1369 | 3854028 | |
BM874316.1 | - | 3 | 358 |