Gene Transcripts

Gene Symbol
NDST4
Entrez Gene ID
64579
Gene Name
N-deacetylase and N-sulfotransferase 4
Gene Aliases
N-HSST, N-HSST 4, NDST-4, NHSST4
Chromosome Location
Chr. 4: 114824122 - 115114098 on Build GRCh38
UniGene ID
Hs.591700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022569.2 NP_072091.1 2 1373
GenBank mRNA AB036429.1 - 2 1373
AK289670.1 - 2 1351

View Full Product Details