Gene Transcripts

Gene Symbol
MYH14
Entrez Gene ID
79784
Gene Name
myosin, heavy chain 14, non-muscle
Gene Aliases
DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
Chromosome Location
Chr. 19: 50188186 - 50310544 on Build GRCh38
UniGene ID
Hs.467142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077186.1 NP_001070654.1 19 2362
NM_001145809.1 NP_001139281.1 20 2461
NM_024729.3 NP_079005.3 18 2338
XM_006723386.3 XP_006723449.1 19 2423
XM_011527320.2 XP_011525622.1 19 2440
XM_011527321.2 XP_011525623.1 18 2416
XM_011527323.2 XP_011525625.1 19 2430
GenBank mRNA AB111886.1 - 8 1020
AB290169.1 - 19 2431
AK125082.1 - 19 2441
AK307783.1 - 19 2344
AY165122.1 - 18 2338
BC018933.2 - 17 2090
FJ041910.1 - 6 624

View Full Product Details