Gene Transcripts

Gene Symbol
ANKRD10
Entrez Gene ID
55608
Gene Name
ankyrin repeat domain 10
Gene Aliases
Chromosome Location
Chr. 13: 110878540 - 110915107 on Build GRCh38
UniGene ID
Hs.525163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017664.3 NP_060134.2 6 2465
NR_104587.1 7 2527
GenBank mRNA AK000100.1 - 6 2426
AK026017.1 - 2 1539
AK094222.1 - 4 2237
AK095763.1 - 4 2137
AK096083.1 - 3 1835
AK123341.1 - 1 2300
BX641022.1 - 1 3461

View Full Product Details