Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018050.2 | NP_060520.2 | 4 | 1209 | |
XM_017019531.1 | XP_016875020.1 | 5 | 1413 | ||
GenBank mRNA | AK001160.1 | - | 5 | 1114 | |
AK023622.1 | - | 4 | 1209 | ||
AK298681.1 | - | 2 | 844 | ||
AY358563.1 | - | 4 | 1176 | ||
BC032998.2 | - | 4 | 1226 | 4824116 |