Gene Transcripts

Gene Symbol
CLSTN1
Entrez Gene ID
22883
Gene Name
calsyntenin 1
Gene Aliases
ALC-ALPHA, CDHR12, CST-1, CSTN1, PIK3CD, XB31alpha, alcalpha1, alcalpha2
Chromosome Location
Chr. 1: 9729021 - 9824526 on Build GRCh38
UniGene ID
Hs.29665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009566.2 NP_001009566.1 2 930
NM_001302883.1 NP_001289812.1 2 930
NM_014944.4 NP_055759.3 2 930
GenBank mRNA AB020718.1 - 2 930
AF438482.1 - 2 896
AK289798.1 - 2 138
AK304813.1 - 2 172
AY753301.1 - 2 137
BC033902.2 - 2 388 5263192

View Full Product Details