Gene Transcripts

Gene Symbol
SKAP2
Entrez Gene ID
8935
Gene Name
src kinase associated phosphoprotein 2
Gene Aliases
PRAP, RA70, SAPS, SCAP2, SKAP-HOM, SKAP55R
Chromosome Location
Chr. 7: 26654772 - 26864743 on Build GRCh38
UniGene ID
Hs.200770
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303468.1 NP_001290397.1 13 2328
NM_003930.4 NP_003921.2 13 2436
GenBank mRNA BC036044.1 - 13 2285

View Full Product Details