Gene Transcripts
- Gene Symbol
- RBM6
- Entrez Gene ID
- 10180
- Gene Name
- RNA binding motif protein 6
- Gene Aliases
- 3G2, DEF-3, DEF3, HLC-11, NY-LU-12, g16
- Chromosome Location
- Chr. 3: 49940038 - 50077252 on Build GRCh38
- UniGene ID
- Hs.696735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001167582.1 |
NP_001161054.1
|
3, 4 |
373 |
|
NM_005777.2 |
NP_005768.1
|
7, 8 |
1886 |
|
XM_005264784.1 |
XP_005264841.1
|
7, 8 |
2161 |
|
XM_005264785.1 |
XP_005264842.1
|
6, 7 |
577 |
|
XM_005264786.1 |
XP_005264843.1
|
5, 6 |
475 |
|
XM_005264787.2 |
XP_005264844.1
|
4, 5 |
1241 |
|
XM_006712916.1 |
XP_006712979.1
|
7, 8 |
1986 |
|
XM_017005496.1 |
XP_016860985.1
|
5, 6 |
1315 |
|
XM_017005497.1 |
XP_016860986.1
|
3, 4 |
210 |
|
XM_017005498.1 |
XP_016860987.1
|
4, 5 |
399 |
|
XM_017005499.1 |
XP_016860988.1
|
9, 10 |
2446 |
|
XM_017005500.1 |
XP_016860989.1
|
8, 9 |
2328 |
|
XM_017005501.1 |
XP_016860990.1
|
8, 9 |
2279 |
|
XM_017005502.1 |
XP_016860991.1
|
3, 4 |
274 |
|
GenBank mRNA |
AF042857.1 |
- |
7, 8 |
1739 |
|
AF069517.1 |
- |
7, 8 |
1842 |
|
AF091264.1 |
- |
7, 8 |
1771 |
|
AK093836.1 |
- |
5, 6 |
418 |
|
AK291719.1 |
- |
7, 8 |
1886 |
|
AK303371.1 |
- |
6, 7 |
1705 |
|
AK308794.1 |
- |
6, 7 |
501 |
|
AL832236.1 |
- |
3, 4 |
322 |
|
BC046643.1 |
- |
3, 4 |
227 |
5753635 |
BX648535.1 |
- |
7, 8 |
2063 |
|
DA068167.1 |
- |
3, 4 |
265 |
|
U50839.1 |
- |
6, 7 |
1672 |
|
View Full Product Details