Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005783.3 | NP_005774.2 | 5 | 1092 | |
GenBank mRNA | AB006679.1 | - | 5 | 1029 | |
AK222616.1 | - | 5 | 1018 | ||
BC005968.1 | - | 5 | 1028 | 4073821 | |
BC022864.1 | - | 5 | 1032 | 4096526 | |
BC070183.1 | - | 5 | 1026 | 4551680 | |
BM553565.1 | - | 408 |