Gene Transcripts

Gene Symbol
RAB3D
Entrez Gene ID
9545
Gene Name
RAB3D, member RAS oncogene family
Gene Aliases
D2-2, GOV, RAB16, RAD3D
Chromosome Location
Chr. 19: 11322046 - 11339668 on Build GRCh38
UniGene ID
Hs.744916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004283.3 NP_004274.1 5 1810
GenBank mRNA AF019226.1 - 1 838
BC016471.1 - 5 1787 3861912

View Full Product Details