Gene Transcripts

Gene Symbol
TROVE2
Entrez Gene ID
6738
Gene Name
TROVE domain family member 2
Gene Aliases
RO60, RORNP, SSA2
Chromosome Location
Chr. 1: 193059422 - 193091777 on Build GRCh38
UniGene ID
Hs.288178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042369.2 NP_001035828.1 2 505
NM_001042370.2 NP_001035829.2 2 505
NM_001173524.1 NP_001166995.1 2 511
NM_001173525.1 NP_001166996.1 2 505
NM_004600.5 NP_004591.2 2 505
XM_006711495.3 XP_006711558.1 2 511
XM_006711496.3 XP_006711559.1 3 687
XM_006711497.3 XP_006711560.1 3 687
XM_017002180.1 XP_016857669.1 2 320
XM_017002181.1 XP_016857670.1 3 615
XM_017002182.1 XP_016857671.1 2 320
GenBank mRNA AK314594.1 - 2 298
AY205314.1 - 2 319
AY205315.1 - 2 319
BC036658.2 - 2 347 4821776
CX761715.1 - 3 645
DA518443.1 - 2 511
J04137.1 - 1 174
M25077.1 - 2 319

View Full Product Details